Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145100.3 | NP_659568.2 | 1 - 2 | 378 | 73 | |
GenBank mRNA | AB041649.1 | - | 1 - 2 | 378 | 73 | |
AK045592.1 | - | 1 - 2 | 379 | 73 | ||
BC058599.1 | - | 1 - 2 | 311 | 6313510 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |