Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172852.3 | NP_766440.2 | 6 - 7 | 671 | 145 | |
GenBank mRNA | AK030670.1 | - | 6 - 7 | 673 | 145 | |
BC117990.1 | - | 6 - 7 | 661 | 40104505 | 145 | |
BC117991.1 | - | 6 - 7 | 608 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |