Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133821.3 | NP_598582.3 | 14 - 15 | 3560 | 86 | |
GenBank mRNA | AB093251.1 | - | 14 - 15 | 2319 | 86 | |
BC024670.1 | - | 2 - 3 | 188 | 86 | ||
BC059254.1 | - | 14 - 15 | 2220 | 6406203 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |