Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133821.3 | NP_598582.3 | 2 - 3 | 1777 | 59 | |
GenBank mRNA | AB093251.1 | - | 2 - 3 | 536 | 59 | |
AK081093.1 | - | 3 - 4 | 842 | 59 | ||
BC059254.1 | - | 2 - 3 | 437 | 6406203 | 59 | |
CN525246.1 | - | 2 - 3 | 243 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |