Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133821.3 | NP_598582.3 | 5 - 6 | 2217 | 75 | |
GenBank mRNA | AB093251.1 | - | 5 - 6 | 976 | 75 | |
BC059254.1 | - | 5 - 6 | 877 | 6406203 | 75 | |
CF535385.1 | - | 2 - 3 | 214 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |