Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022312.3 | NP_071707.2 | 8 - 9 | 2386 | 89 | |
GenBank mRNA | AK039390.1 | - | 8 - 9 | 2387 | 89 | |
AK078229.1 | - | 1 - 2 | 64 | 89 | ||
BC132392.1 | - | 8 - 9 | 2020 | 40130669 | 89 | |
BC138043.1 | - | 8 - 9 | 2020 | 8861062 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |