Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009786.2 | NP_033916.1 | 4 - 5 | 666 | 88 | |
GenBank mRNA | AK013924.1 | - | 4 - 5 | 597 | 88 | |
AK089043.1 | - | 4 - 5 | 520 | 88 | ||
AK212050.1 | - | 2 - 3 | 130 | 88 | ||
AK215280.1 | - | 2 - 3 | 130 | 88 | ||
BC025948.1 | - | 4 - 5 | 551 | 5150537 | 88 | |
U97327.1 | - | 4 - 5 | 496 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |