Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026500.3 | NP_080776.1 | 6 - 7 | 1636 | 65 | |
GenBank mRNA | AK004833.1 | - | 6 - 7 | 1637 | 65 | |
AK133731.1 | - | 6 - 7 | 1598 | 65 | ||
BC023840.1 | - | 6 - 7 | 1563 | 5344890 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |