Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145456.2 | NP_663431.2 | 2 - 3 | 1025 | 83 | |
GenBank mRNA | AK122528.1 | - | 2 - 3 | 440 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |