Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008473.2 | NP_032499.2 | 5 - 6 | 1217 | 64 | |
GenBank mRNA | AK019521.1 | - | 5 - 6 | 1218 | 64 | |
BC117842.1 | - | 5 - 6 | 1184 | 40093577 | 64 | |
BC117843.1 | - | 5 - 6 | 1184 | 40093580 | 64 | |
M10937.1 | - | 5 - 6 | 1225 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |