Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172564.3 | NP_766152.2 | 10 - 11 | 2017 | 69 | |
GenBank mRNA | AK036834.1 | - | 10 - 11 | 2014 | 69 | |
AK039855.1 | - | 5 - 6 | 643 | 69 | ||
AK170638.1 | - | 4 - 5 | 389 | 69 | ||
BC055820.1 | - | 10 - 11 | 1999 | 4209161 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |