Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026313.1 | NP_080589.1 | 2 - 3 | 288 | 157 | |
GenBank mRNA | AK014362.1 | - | 2 - 3 | 281 | 157 | |
AK019464.1 | - | 2 - 3 | 341 | 157 | ||
AK078090.1 | - | 2 - 3 | 335 | 157 | ||
AK151228.1 | - | 2 - 3 | 336 | 157 | ||
AK151839.1 | - | 2 - 3 | 336 | 157 | ||
BC009092.1 | - | 2 - 3 | 288 | 3157532 | 157 | |
EF410121.1 | - | 320 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |