Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153518.1 | NP_705738.1 | 1 - 2 | 316 | 62 | |
GenBank mRNA | AF454432.1 | - | 1 - 2 | 316 | 62 | |
AK077174.1 | - | 1 - 2 | 350 | 62 | ||
BC048073.1 | - | 1 - 2 | 336 | 6312435 | 62 | |
BC056947.1 | - | 1 - 2 | 312 | 62 | ||
BC060155.1 | - | 1 - 2 | 312 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |