Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011711.1 | NP_035841.1 | 2 - 3 | 370 | 134 | |
GenBank mRNA | AK129473.1 | - | 2 - 3 | 289 | 134 | |
AK139711.1 | - | 2 - 3 | 353 | 134 | ||
AK143308.1 | - | 2 - 3 | 269 | 134 | ||
AK155331.1 | - | 2 - 3 | 387 | 134 | ||
AK163361.1 | - | 2 - 3 | 354 | 134 | ||
AK170744.1 | - | 2 - 3 | 353 | 134 | ||
BC023797.1 | - | 1 - 2 | 90 | 134 | ||
BC085094.1 | - | 2 - 3 | 267 | 134 | ||
BC131961.1 | - | 2 - 3 | 246 | 40130238 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |