Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007405.2 | NP_031431.2 | 11 - 12 | 2137 | 63 | |
GenBank mRNA | AK147516.1 | - | 11 - 12 | 2137 | 63 | |
BC145101.1 | - | 10 - 11 | 1970 | 63 | ||
M93422.1 | - | 10 - 11 | 2069 | 63 | ||
M96653.1 | - | 10 - 11 | 1959 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |