Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100014087.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014087.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014087.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014084.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014084.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014084.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014088.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014088.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014088.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014093.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014093.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014093.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014082.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014082.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014082.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014083.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014083.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014083.mm.1 |
TC1100001530.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100014091.mm.1 |
NM_001081434.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014091.mm.1 |
NM_001081434.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100014091.mm.1 |
TC1100001530.mm.1 |
MTA TC |