Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142580.1 | NP_001136052.1 | 10 - 11 | 862 | 103 | |
NM_001142581.1 | NP_001136053.1 | 9 - 10 | 805 | 103 | ||
NM_134044.3 | NP_598805.2 | 9 - 10 | 819 | 103 | ||
GenBank mRNA | AK038165.1 | - | 10 - 11 | 830 | 103 | |
AK046322.1 | - | 10 - 11 | 833 | 103 | ||
AK049643.1 | - | 10 - 11 | 809 | 103 | ||
AK079055.1 | - | 10 - 11 | 809 | 103 | ||
AK082261.1 | - | 10 - 11 | 790 | 103 | ||
AK145043.1 | - | 845 | 103 | |||
BC016646.1 | - | 9 - 10 | 728 | 2651233 | 103 | |
BC023716.1 | - | 9 - 10 | 830 | 5346489 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |