Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252556.1 | NP_001239485.1 | 1 - 2 | 142 | 58 | |
NM_010363.4 | NP_034493.1 | 1 - 2 | 142 | 58 | ||
GenBank mRNA | AF093418.1 | - | 1 - 2 | 91 | 58 | |
AK002398.1 | - | 1 - 2 | 99 | 58 | ||
AK012796.1 | - | 1 - 2 | 142 | 58 | ||
AK075927.1 | - | 1 - 2 | 96 | 58 | ||
BC031777.1 | - | 1 - 2 | 131 | 4168439 | 58 | |
BY072664.1 | - | 1 - 2 | 143 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |