Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164785.1 | NP_001158257.1 | 11 - 12 | 1869 | 96 | |
NM_177431.4 | NP_803180.3 | 11 - 12 | 1869 | 96 | ||
GenBank mRNA | AJ512753.3 | - | 11 - 12 | 1638 | 96 | |
AY189815.1 | - | 11 - 12 | 1777 | 96 | ||
AY189816.1 | - | 11 - 12 | 1777 | 96 | ||
BC121792.1 | - | 11 - 12 | 1708 | 40099393 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |