Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00018924.mm.1 |
NM_001024141.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018924.mm.1 |
NM_001024141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018924.mm.1 |
TC0X00003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018926.mm.1 |
NM_001024141.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018926.mm.1 |
NM_001024141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018926.mm.1 |
TC0X00003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018925.mm.1 |
NM_001024141.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018925.mm.1 |
NM_001024141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018925.mm.1 |
TC0X00003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018927.mm.1 |
NM_001024141.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018927.mm.1 |
NM_001024141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018927.mm.1 |
TC0X00003019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00018928.mm.1 |
NM_001024141.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018928.mm.1 |
NM_001024141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00018928.mm.1 |
TC0X00003019.mm.1 |
MTA TC |