Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130888.1 | NP_570958.1 | 18 - 19 | 1516 | 66 | |
GenBank mRNA | AB178901.1 | - | 19 - 20 | 1655 | 66 | |
AJ305317.1 | - | 14 - 15 | 1077 | 66 | ||
AJ305318.1 | - | 15 - 16 | 1185 | 66 | ||
AY260550.1 | - | 18 - 19 | 1516 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |