Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134091.2 | NP_598852.2 | 19 - 20 | 2182 | 79 | |
GenBank mRNA | AK142314.1 | - | 19 - 20 | 2150 | 79 | |
AK170806.1 | - | 19 - 20 | 2149 | 79 | ||
AY382616.1 | - | 19 - 20 | 2136 | 79 | ||
BC018197.1 | - | 19 - 20 | 2142 | 4190407 | 79 | |
BC024122.1 | - | 19 - 20 | 2142 | 5097970 | 79 | |
BC025561.1 | - | 19 - 20 | 2120 | 5322980 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |