Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033217.4 | NP_001028389.1 | 3 - 4 | 499 | 147 | |
GenBank mRNA | AK147883.1 | - | 3 - 4 | 512 | 147 | |
AK153711.1 | - | 3 - 4 | 500 | 147 | ||
BC117892.1 | - | 3 - 4 | 471 | 40096843 | 147 | |
BC117893.1 | - | 3 - 4 | 471 | 40096845 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |