Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159647.1 | NP_001153119.1 | 4 - 5 | 610 | 78 | |
NM_001159648.1 | NP_001153120.1 | 4 - 5 | 464 | 78 | ||
NM_007727.2 | NP_031753.1 | 4 - 5 | 513 | 78 | ||
GenBank mRNA | AK138969.1 | - | 4 - 5 | 465 | 78 | |
BC066864.1 | - | 4 - 5 | 536 | 30099512 | 78 | |
X14943.1 | - | 4 - 5 | 483 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |