Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172594.2 | NP_766182.2 | 4 - 5 | 503 | 101 | |
GenBank mRNA | AK042066.1 | - | 4 - 5 | 764 | 101 | |
AK042497.1 | - | 4 - 5 | 766 | 101 | ||
AK049530.1 | - | 4 - 5 | 576 | 101 | ||
AK087450.1 | - | 4 - 5 | 842 | 101 | ||
BC020331.1 | - | 4 - 5 | 596 | 101 | ||
BC037686.1 | - | 4 - 5 | 579 | 101 | ||
BC057112.1 | - | 4 - 5 | 576 | 6417650 | 101 | |
BC082319.1 | - | 4 - 5 | 566 | 6417162 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |