Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253813.1 | NP_001240742.1 | 17 - 18 | 2217 | 79 | |
NM_175344.4 | NP_780553.2 | 16 - 17 | 2154 | 79 | ||
GenBank mRNA | AK089300.1 | - | 17 - 18 | 2290 | 79 | |
AK159411.1 | - | 17 - 18 | 2166 | 79 | ||
BC060732.1 | - | 16 - 17 | 2154 | 6409332 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |