Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177323.2 | NP_001170794.1 | 8 - 9 | 1376 | 68 | |
NM_001177324.1 | NP_001170795.1 | 8 - 9 | 1379 | 68 | ||
NM_001253706.1 | NP_001240635.1 | 8 - 9 | 1379 | 68 | ||
NM_019942.5 | NP_064326.2 | 8 - 9 | 1379 | 68 | ||
GenBank mRNA | AB023622.1 | - | 8 - 9 | 1143 | 68 | |
AK012858.1 | - | 9 - 10 | 1170 | 68 | ||
AK048455.1 | - | 8 - 9 | 1201 | 68 | ||
AK088614.1 | - | 8 - 9 | 1142 | 68 | ||
AK160884.1 | - | 8 - 9 | 1173 | 68 | ||
AK172894.1 | - | 8 - 9 | 1160 | 68 | ||
BC010489.1 | - | 8 - 9 | 1165 | 4168214 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |