Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019668.3 | NP_062642.1 | 1 - 2 | 160 | 53 | |
GenBank mRNA | AF089812.1 | - | 1 - 2 | 168 | 53 | |
AF383148.1 | - | 1 - 2 | 168 | 53 | ||
AK133723.1 | - | 1 - 2 | 169 | 53 | ||
AK150417.1 | - | 1 - 2 | 143 | 53 | ||
AK153327.1 | - | 1 - 2 | 143 | 53 | ||
AK160527.1 | - | 1 - 2 | 155 | 53 | ||
AK160538.1 | - | 1 - 2 | 169 | 53 | ||
AK160545.1 | - | 1 - 2 | 167 | 53 | ||
AK163966.1 | - | 1 - 2 | 195 | 53 | ||
AK164074.1 | - | 1 - 2 | 169 | 53 | ||
BC026053.1 | - | 1 - 2 | 158 | 4012075 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |