Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081402.1 | NP_001074871.1 | 7 - 8 | 762 | 99 | |
GenBank mRNA | AK159714.1 | - | 7 - 8 | 744 | 99 | |
AK160933.1 | - | 7 - 8 | 697 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |