Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081402.1 | NP_001074871.1 | 9 - 10 | 993 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |