Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029934.1 | NP_001025105.1 | 21 - 22 | 2611 | 93 | |
GenBank mRNA | AK148221.1 | - | 21 - 22 | 2611 | 93 | |
BC058994.1 | - | 2 - 3 | 152 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |