Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159538.1 | NP_001153010.1 | 14 - 15 | 1789 | 83 | |
NM_013710.4 | NP_038738.2 | 14 - 15 | 1786 | 83 | ||
GenBank mRNA | AF017368.1 | - | 14 - 15 | 1728 | 83 | |
AK042260.1 | - | 14 - 15 | 1787 | 83 | ||
AK156151.1 | - | 14 - 15 | 1786 | 83 | ||
BC021845.1 | - | 11 - 12 | 1241 | 4016697 | 83 | |
DQ344523.1 | - | 14 - 15 | 1610 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |