Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198647.1 | NP_941049.1 | 6 - 7 | 939 | 62 | |
GenBank mRNA | BC045600.1 | - | 6 - 7 | 939 | 4489206 | 62 |
BC060066.1 | - | 1 - 2 | 211 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |