Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178309.2 | NP_840094.1 | 9 - 10 | 1461 | 88 | |
GenBank mRNA | AK033205.1 | - | 9 - 10 | 1432 | 88 | |
AK051878.1 | - | 9 - 10 | 1462 | 88 | ||
AK145709.1 | - | 9 - 10 | 1462 | 88 | ||
AK169440.1 | - | 9 - 10 | 1408 | 88 | ||
AK169823.1 | - | 8 - 9 | 1198 | 88 | ||
BC094252.1 | - | 9 - 10 | 1410 | 6401109 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |