Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081020.1 | NP_001074489.1 | 7 - 8 | 1470 | 80 | |
GenBank mRNA | AK030315.1 | - | 7 - 8 | 1623 | 80 | |
AK044528.1 | - | 7 - 8 | 1473 | 80 | ||
BC172116.1 | - | 7 - 8 | 1373 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |