Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289496.1 | NP_001276425.1 | 6 - 7 | 1015 | 122 | |
NM_001289498.1 | NP_001276427.1 | 6 - 7 | 1010 | 122 | ||
NM_178748.6 | NP_848863.1 | 6 - 7 | 1015 | 122 | ||
GenBank mRNA | AK031136.1 | - | 6 - 7 | 924 | 122 | |
AK037223.1 | - | 6 - 7 | 917 | 122 | ||
AK041546.1 | - | 6 - 7 | 896 | 122 | ||
AK087659.1 | - | 6 - 7 | 895 | 122 | ||
AK158909.1 | - | 6 - 7 | 1011 | 122 | ||
BC095994.1 | - | 6 - 7 | 745 | 6467602 | 122 | |
BC150710.1 | - | 6 - 7 | 735 | 9087621 | 122 | |
DQ223720.1 | - | 6 - 7 | 719 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |