Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030168.3 | NP_084444.3 | 27 - 28 | 2713 | 72 | |
GenBank mRNA | AK173326.1 | - | 3 - 4 | 218 | 72 | |
AY497009.1 | - | 27 - 28 | 2691 | 72 | ||
AY540053.1 | - | 27 - 28 | 2711 | 72 | ||
BC022906.1 | - | 22 - 23 | 2259 | 72 | ||
BC120903.1 | - | 23 - 24 | 2352 | 40086992 | 72 | |
BC120904.1 | - | 27 - 28 | 2711 | 40086995 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |