Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_001172147.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_001172147.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_001172148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_001172148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_153586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
NM_153586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019267.mm.1 |
TC0X00003067.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_001172147.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_001172147.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_001172148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_001172148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_153586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
NM_153586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019275.mm.1 |
TC0X00003067.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_001172147.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_001172147.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_001172148.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_001172148.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_153586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
NM_153586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019270.mm.1 |
TC0X00003067.mm.1 |
MTA TC |