Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172147.1 | NP_001165618.1 | 5 - 6 | 699 | 139 | |
NM_001172148.1 | NP_001165619.1 | 4 - 5 | 617 | 139 | ||
NM_153586.2 | NP_705814.1 | 4 - 5 | 617 | 139 | ||
GenBank mRNA | AK040213.1 | - | 4 - 5 | 557 | 139 | |
AK080041.1 | - | 2 - 3 | 383 | 139 | ||
AK084514.1 | - | 4 - 5 | 552 | 139 | ||
AK084629.1 | - | 4 - 5 | 552 | 139 | ||
AK163147.1 | - | 3 - 4 | 536 | 139 | ||
AK164496.1 | - | 5 - 6 | 647 | 139 | ||
AK166005.1 | - | 4 - 5 | 552 | 139 | ||
AK166229.1 | - | 565 | 139 | |||
BC037024.1 | - | 4 - 5 | 584 | 5368185 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |