Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00003428.mm.1 |
NM_172780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003428.mm.1 |
NM_172780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003428.mm.1 |
TC0X00000522.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003432.mm.1 |
NM_172780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003432.mm.1 |
NM_172780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003432.mm.1 |
TC0X00000522.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003429.mm.1 |
NM_172780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003429.mm.1 |
NM_172780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003429.mm.1 |
TC0X00000522.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003430.mm.1 |
NM_172780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003430.mm.1 |
NM_172780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003430.mm.1 |
TC0X00000522.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00003431.mm.1 |
NM_172780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003431.mm.1 |
NM_172780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00003431.mm.1 |
TC0X00000522.mm.1 |
MTA TC |