Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145885.1 | NP_001139357.1 | 15 - 16 | 1219 | 90 | |
NM_010029.2 | NP_034159.1 | 14 - 15 | 1141 | 90 | ||
GenBank mRNA | AK014844.1 | - | 14 - 15 | 1132 | 90 | |
AK133366.1 | - | 15 - 16 | 1212 | 90 | ||
BC137601.1 | - | 14 - 15 | 1195 | 8860620 | 90 | |
BC144760.1 | - | 14 - 15 | 1195 | 9053308 | 90 | |
JQ923486.1 | - | 14 - 15 | 1169 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |