Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010897.2 | NP_035027.1 | 42 - 43 | 6547 | 74 | |
GenBank mRNA | AK158728.1 | - | 5 - 6 | 1052 | 74 | |
L10370.1 | - | 41 - 42 | 6392 | 74 | ||
X54924.1 | - | 3 - 4 | 592 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |