Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175501.2 | NP_780710.2 | 3 - 4 | 889 | 69 | |
GenBank mRNA | AJ537452.1 | - | 3 - 4 | 648 | 69 | |
AK048612.1 | - | 3 - 4 | 852 | 69 | ||
BC151248.1 | - | 3 - 4 | 652 | 69 | ||
BC152338.1 | - | 3 - 4 | 652 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |