Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018873.2 | NP_061361.2 | 15 - 16 | 2967 | 64 | |
GenBank mRNA | AF040944.1 | - | 15 - 16 | 2969 | 64 | |
AK129422.1 | - | 10 - 11 | 2124 | 64 | ||
U59873.1 | - | 15 - 16 | 2968 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |