Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139311.2 | NP_647472.2 | 10 - 11 | 1777 | 85 | |
GenBank mRNA | AK143477.1 | - | 6 - 7 | 1220 | 85 | |
AY050217.1 | - | 10 - 11 | 1924 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |