Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029430.1 | NP_083706.1 | 6 - 7 | 843 | 91 | |
GenBank mRNA | AK015027.1 | - | 6 - 7 | 815 | 91 | |
BC132384.1 | - | 6 - 7 | 843 | 91 | ||
BC145852.1 | - | 6 - 7 | 843 | 40131155 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |