Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033337.4 | NP_001028509.2 | 6 - 7 | 698 | 68 | |
GenBank mRNA | AK078848.1 | - | 6 - 7 | 659 | 68 | |
AK133636.1 | - | 5 - 6 | 406 | 68 | ||
BC024550.1 | - | 6 - 7 | 648 | 68 | ||
BC030849.1 | - | 6 - 7 | 661 | 68 | ||
BC132621.1 | - | 6 - 7 | 683 | 40130898 | 68 | |
BC132623.1 | - | 6 - 7 | 683 | 40130900 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |