Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038619.1 | NP_001033708.1 | 14 - 15 | 1867 | 82 | |
NM_172646.2 | NP_766234.1 | 14 - 15 | 1871 | 82 | ||
GenBank mRNA | AK036199.1 | - | 14 - 15 | 1832 | 82 | |
AK049724.1 | - | 14 - 15 | 1872 | 82 | ||
AK082698.1 | - | 14 - 15 | 1811 | 82 | ||
AK147678.1 | - | 14 - 15 | 1811 | 82 | ||
BC004050.1 | - | 3 - 4 | 239 | 82 | ||
BC024584.1 | - | 3 - 4 | 271 | 82 | ||
BC141143.1 | - | 14 - 15 | 1715 | 9055755 | 82 | |
BC141144.1 | - | 14 - 15 | 1715 | 9055756 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |