Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100027708.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027708.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027708.mm.1 |
NM_172646.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027708.mm.1 |
NM_172646.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027708.mm.1 |
TC0100003382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027709.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027709.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027709.mm.1 |
NM_172646.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027709.mm.1 |
NM_172646.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027709.mm.1 |
TC0100003382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027702.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027702.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027702.mm.1 |
TC0100003382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027701.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027701.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027701.mm.1 |
TC0100003382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027705.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027705.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027705.mm.1 |
NM_172646.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027705.mm.1 |
NM_172646.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027705.mm.1 |
TC0100003382.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027704.mm.1 |
NM_001038619.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027704.mm.1 |
NM_001038619.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027704.mm.1 |
NM_172646.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027704.mm.1 |
NM_172646.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027704.mm.1 |
TC0100003382.mm.1 |
MTA TC |