Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027185.3 | NP_081461.2 | 3 - 4 | 490 | 64 | |
GenBank mRNA | AF329497.1 | - | 3 - 4 | 425 | 64 | |
AK088460.1 | - | 3 - 4 | 439 | 64 | ||
AY241695.1 | - | 3 - 4 | 425 | 64 | ||
BC120500.1 | - | 3 - 4 | 490 | 40129423 | 64 | |
BC131953.1 | - | 3 - 4 | 490 | 40130230 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |